Merck
LEF-1 Regulates Tyrosinase Gene Transcription In Vitro.
PloS one (2015-11-19)
Xueping Wang, Yalan Liu, Hongsheng Chen, Lingyun Mei, Chufeng He, Lu Jiang, Zhijie Niu, Jie Sun, Hunjin Luo, Jiada Li, Yong Feng
ABSTRACT
TYR, DCT and MITF are three important genes involved in maintaining the mature phenotype and producing melanin; they therefore participate in neural crest cell development into melanocytes. Previous studies have revealed that the Wnt signaling factor lymphoid enhancer-binding factor (LEF-1) can enhance DCT and MITF gene expression. However, whether LEF-1 also affects TYR gene expression remains unclear. In the present study, we found that LEF-1 regulated TYR transcription in vitro. LEF-1 overexpression increased TYR gene promoter activity, whereas LEF-1 knockdown by RNA interference significantly decreased TYR expression. Moreover, the core GTTTGAT sequence (-56 to -50) within the TYR promoter is essential for the effect of LEF-1 on TYR expression, and chromatin immunoprecipitation (ChIP) assay indicated that endogenous LEF-1 interacts with the TYR promoter. In addition, we observed a synergistic transactivation of the TYR promoter by LEF-1 and MITF. These data suggest that Wnt signaling plays an important role in regulating melanocyte development and differentiation.
Sigma-Aldrich
Monoclonal Anti-Microphthalmia antibody produced in mouse, clone C5, purified immunoglobulin
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)