Merck
  • The indirect pathway of the nucleus accumbens shell amplifies neuropathic pain.
The indirect pathway of the nucleus accumbens shell amplifies neuropathic pain.
Nature neuroscience (2015-12-23)
Wenjie Ren, Maria Virginia Centeno, Sara Berger, Ying Wu, Xiaodong Na, Xianguo Liu, Jyothisri Kondapalli, A Vania Apkarian, Marco Martina, D James Surmeier
ABSTRACT
We examined adaptations in nucleus accumbens (NAc) neurons in mouse and rat peripheral nerve injury models of neuropathic pain. Injury selectively increased excitability of NAc shell indirect pathway spiny projection neurons (iSPNs) and altered their synaptic connectivity. Moreover, injury-induced tactile allodynia was reversed by inhibiting and exacerbated by exciting iSPNs, indicating that they not only participated in the central representation of pain, but gated activity in ascending nociceptive pathways.
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
PSEM89S Hydrochloride salt, ≥98% (HPLC)