Merck
Modular Synthetic Inverters from Zinc Finger Proteins and Small RNAs.
PloS one (2016-02-18)
Justin Hsia, William J Holtz, Michel M Maharbiz, Murat Arcak, Jay D Keasling
ABSTRACT
Synthetic zinc finger proteins (ZFPs) can be created to target promoter DNA sequences, repressing transcription. The binding of small RNA (sRNA) to ZFP mRNA creates an ultrasensitive response to generate higher effective Hill coefficients. Here we combined three "off the shelf" ZFPs and three sRNAs to create new modular inverters in E. coli and quantify their behavior using induction fold. We found a general ordering of the effects of the ZFPs and sRNAs on induction fold that mostly held true when combining these parts. We then attempted to construct a ring oscillator using our new inverters. Our chosen parts performed insufficiently to create oscillations, but we include future directions for improvement upon our work presented here.
Product Description
AeraSealā„¢ film, sterile