Merck
  • SF Treg cells transcribing high levels of Bcl-2 and microRNA-21 demonstrate limited apoptosis in RA.
SF Treg cells transcribing high levels of Bcl-2 and microRNA-21 demonstrate limited apoptosis in RA.
Rheumatology (Oxford, England) (2014-10-24)
Kornelis S M van der Geest, Katarzyna Smigielska-Czepiel, Ji-Ah Park, Wayel H Abdulahad, Hye-Won Kim, Bart-Jan Kroesen, Anke van den Berg, Annemieke M H Boots, Eun-Bong Lee, Elisabeth Brouwer
ABSTRACT
The aim of this study was to investigate the turnover of Treg cells in the SF of RA patients. Treg cells were enumerated in peripheral blood and SF of RA patients and analysed by flow cytometry for expression of the proliferation marker Ki-67 and binding of the apoptosis marker annexin V. Sorted Treg cells of RA patients were analysed for expression of anti-apoptotic regulators Bcl-2 and microRNA-21 (miR-21) by RT-PCR. Treg cells displaying a memory phenotype were abundant in the SF of RA patients. SF Treg cells more frequently expressed the proliferation marker Ki-67 than conventional T cells. Only few SF Treg cells were apoptotic, as indicated by limited annexin V staining of these cells. SF Treg cells displayed high transcription levels of Bcl-2 and miR-21 in comparison with SF conventional T cells and peripheral blood Treg cells. Treg cells with a memory phenotype accumulate in the SF of RA patients. These Treg cells have a high proliferative activity and demonstrate little apoptosis. The latter finding could be explained by high transcription of Bcl-2 and miR-21 in SF Treg cells.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required