Merck
  • Intravenous peramivir inhibits viral replication, and leads to bacterial clearance and prevention of mortality during murine bacterial co-infection caused by influenza A(H1N1)pdm09 virus and Streptococcus pneumoniae.
Intravenous peramivir inhibits viral replication, and leads to bacterial clearance and prevention of mortality during murine bacterial co-infection caused by influenza A(H1N1)pdm09 virus and Streptococcus pneumoniae.
Antiviral research (2015-03-11)
Motoyasu Onishi, Mitsutaka Kitano, Keiichi Taniguchi, Tomoyuki Homma, Masanori Kobayashi, Tomokazu Yoshinaga, Akira Naito, Akihiko Sato
ABSTRACT
Influenza virus infection increases susceptibility to bacterial infection and mortality in humans. Although the efficacy of approved intravenous peramivir, a neuraminidase (NA) inhibitor, against influenza virus infection has been reported, its efficacy against bacterial co-infection, which occurs during the period of viral shedding, was not fully investigated. To further understand the significance of treatment with peramivir, we assessed the efficacy of peramivir against a bacterial co-infection model in mice caused by clinically isolated influenza A(H1N1)pdm09 virus and Streptococcus pneumoniae. Mice were infected with influenza A(H1N1)pdm09. Peramivir was intravenously administered after the viral infection. At 2days post viral infection, the mice were infected with S. pneumoniae. Peramivir efficacy was measured by the survival rates and viral titers, bacterial titers, or proinflammatory cytokine concentrations in lung homogenates. Peramivir treatment reduced the mortality of mice infected with influenza virus and S. pneumoniae. The survival rate in the peramivir-treated group was significantly higher than that in the oseltamivir-treated group. Viral titers and proinflammatory cytokine responses in the peramivir-treated group were significantly lower than those in the oseltamivir-treated group until at 2days post viral infection. Bacterial titer was significantly lower in the peramivir-treated group than in the oseltamivir-treated group at 4days post viral infection. These results demonstrated that peramivir inhibits viral replication, consequently leading to bacterial clearance and prevention of mortality during severe murine bacterial co-infection, which occurs during the period of viral shedding, with the efficacy of peramivir being superior to that of oseltamivir.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture