Merck
  • SOD1 protein aggregates stimulate macropinocytosis in neurons to facilitate their propagation.
SOD1 protein aggregates stimulate macropinocytosis in neurons to facilitate their propagation.
Molecular neurodegeneration (2015-11-02)
Rafaa Zeineddine, Jay F Pundavela, Lisa Corcoran, Elise M Stewart, Dzung Do-Ha, Monique Bax, Gilles Guillemin, Kara L Vine, Danny M Hatters, Heath Ecroyd, Christopher M Dobson, Bradley J Turner, Lezanne Ooi, Mark R Wilson, Neil R Cashman, Justin J Yerbury
ABSTRACT
Amyotrophic Lateral Sclerosis is characterized by a focal onset of symptoms followed by a progressive spread of pathology that has been likened to transmission of infectious prions. Cell-to-cell transmission of SOD1 protein aggregates is dependent on fluid-phase endocytosis pathways, although the precise molecular mechanisms remain to be elucidated. We demonstrate in this paper that SOD1 aggregates interact with the cell surface triggering activation of Rac1 and subsequent membrane ruffling permitting aggregate uptake via stimulated macropinocytosis. In addition, other protein aggregates, including those associated with neurodegenerative diseases (TDP-43, Httex146Q, α-synuclein) also trigger membrane ruffling to gain entry into the cell. Aggregates are able to rupture unstructured macropinosomes to enter the cytosol allowing propagation of aggregation to proceed. Thus, we conclude that in addition to basic proteostasis mechanisms, pathways involved in the activation of macropinocytosis are key determinants in the spread of pathology in these misfolding diseases.
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications