Merck
  • Adjunct prednisone therapy for patients with community-acquired pneumonia: a multicentre, double-blind, randomised, placebo-controlled trial.
Adjunct prednisone therapy for patients with community-acquired pneumonia: a multicentre, double-blind, randomised, placebo-controlled trial.
Lancet (London, England) (2015-01-23)
Claudine Angela Blum, Nicole Nigro, Matthias Briel, Philipp Schuetz, Elke Ullmer, Isabelle Suter-Widmer, Bettina Winzeler, Roland Bingisser, Hanno Elsaesser, Daniel Drozdov, Birsen Arici, Sandrine Andrea Urwyler, Julie Refardt, Philip Tarr, Sebastian Wirz, Robert Thomann, Christine Baumgartner, Hervé Duplain, Dieter Burki, Werner Zimmerli, Nicolas Rodondi, Beat Mueller, Mirjam Christ-Crain
ABSTRACT
Clinical trials yielded conflicting data about the benefit of adding systemic corticosteroids for treatment of community-acquired pneumonia. We assessed whether short-term corticosteroid treatment reduces time to clinical stability in patients admitted to hospital for community-acquired pneumonia. In this double-blind, multicentre, randomised, placebo-controlled trial, we recruited patients aged 18 years or older with community-acquired pneumonia from seven tertiary care hospitals in Switzerland within 24 h of presentation. Patients were randomly assigned (1:1 ratio) to receive either prednisone 50 mg daily for 7 days or placebo. The computer-generated randomisation was done with variable block sizes of four to six and stratified by study centre. The primary endpoint was time to clinical stability defined as time (days) until stable vital signs for at least 24 h, and analysed by intention to treat. This trial is registered with ClinicalTrials.gov, number NCT00973154. From Dec 1, 2009, to May 21, 2014, of 2911 patients assessed for eligibility, 785 patients were randomly assigned to either the prednisone group (n=392) or the placebo group (n=393). Median time to clinical stability was shorter in the prednisone group (3·0 days, IQR 2·5-3·4) than in the placebo group (4·4 days, 4·0-5·0; hazard ratio [HR] 1·33, 95% CI 1·15-1·50, p<0·0001). Pneumonia-associated complications until day 30 did not differ between groups (11 [3%] in the prednisone group and 22 [6%] in the placebo group; odds ratio [OR] 0·49 [95% CI 0·23-1·02]; p=0·056). The prednisone group had a higher incidence of in-hospital hyperglycaemia needing insulin treatment (76 [19%] vs 43 [11%]; OR 1·96, 95% CI 1·31-2·93, p=0·0010). Other adverse events compatible with corticosteroid use were rare and similar in both groups. Prednisone treatment for 7 days in patients with community-acquired pneumonia admitted to hospital shortens time to clinical stability without an increase in complications. This finding is relevant from a patient perspective and an important determinant of hospital costs and efficiency. Swiss National Science Foundation, Viollier AG, Nora van Meeuwen Haefliger Stiftung, Julia und Gottfried Bangerter-Rhyner Stiftung.
Product Description
Prednisone, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prednisone, ≥98%
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard