Merck
  • Au nanomatryoshkas as efficient near-infrared photothermal transducers for cancer treatment: benchmarking against nanoshells.
Au nanomatryoshkas as efficient near-infrared photothermal transducers for cancer treatment: benchmarking against nanoshells.
ACS nano (2014-06-04)
Ciceron Ayala-Orozco, Cordula Urban, Mark W Knight, Alexander Skyrme Urban, Oara Neumann, Sandra W Bishnoi, Shaunak Mukherjee, Amanda M Goodman, Heather Charron, Tamika Mitchell, Martin Shea, Ronita Roy, Sarmistha Nanda, Rachel Schiff, Naomi J Halas, Amit Joshi
ABSTRACT
Au nanoparticles with plasmon resonances in the near-infrared (NIR) region of the spectrum efficiently convert light into heat, a property useful for the photothermal ablation of cancerous tumors subsequent to nanoparticle uptake at the tumor site. A critical aspect of this process is nanoparticle size, which influences both tumor uptake and photothermal efficiency. Here, we report a direct comparative study of ∼90 nm diameter Au nanomatryoshkas (Au/SiO2/Au) and ∼150 nm diameter Au nanoshells for photothermal therapeutic efficacy in highly aggressive triple negative breast cancer (TNBC) tumors in mice. Au nanomatryoshkas are strong light absorbers with 77% absorption efficiency, while the nanoshells are weaker absorbers with only 15% absorption efficiency. After an intravenous injection of Au nanomatryoshkas followed by a single NIR laser dose of 2 W/cm(2) for 5 min, 83% of the TNBC tumor-bearing mice appeared healthy and tumor free >60 days later, while only 33% of mice treated with nanoshells survived the same period. The smaller size and larger absorption cross section of Au nanomatryoshkas combine to make this nanoparticle more effective than Au nanoshells for photothermal cancer therapy.
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis