Merck
  • Autistic-like syndrome in mu opioid receptor null mice is relieved by facilitated mGluR4 activity.
Autistic-like syndrome in mu opioid receptor null mice is relieved by facilitated mGluR4 activity.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2014-03-13)
Jérôme A J Becker, Daniel Clesse, Coralie Spiegelhalter, Yannick Schwab, Julie Le Merrer, Brigitte L Kieffer
ABSTRACT
The etiology of Autism Spectrum Disorders (ASDs) remains largely unknown. Identifying vulnerability genes for autism represents a major challenge in the field and allows the development of animal models for translational research. Mice lacking the mu opioid receptor gene (Oprm1(-/-)) were recently proposed as a monogenic mouse model of autism, based on severe deficits in social behavior and communication skills. We confirm this hypothesis by showing that adult Oprm1(-/-) animals recapitulate core and multiple comorbid behavioral symptoms of autism and also display anatomical, neurochemical, and genetic landmarks of the disease. Chronic facilitation of mGluR4 signaling, which we identified as a novel pharmacological target in ASDs in these mice, was more efficient in alleviating behavioral deficits than the reference molecule risperidone. Altogether, our data provide first evidence that disrupted mu opioid receptor signaling is sufficient to trigger a comprehensive autistic syndrome, maybe through blunted social reward processes, and this mouse model opens promising avenues for therapeutic innovation.
Product Description
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Risperidone, European Pharmacopoeia (EP) Reference Standard