Merck
  • Proprotein convertase subtilisin kexin type 9 promotes intestinal overproduction of triglyceride-rich apolipoprotein B lipoproteins through both low-density lipoprotein receptor-dependent and -independent mechanisms.
Proprotein convertase subtilisin kexin type 9 promotes intestinal overproduction of triglyceride-rich apolipoprotein B lipoproteins through both low-density lipoprotein receptor-dependent and -independent mechanisms.
Circulation (2014-07-30)
Shirya Rashid, Hagai Tavori, Patrick E Brown, MacRae F Linton, Jane He, Ilaria Giunzioni, Sergio Fazio
ABSTRACT
Proprotein convertase subtilisin kexin type 9 (PCSK9) promotes the degradation of the low-density lipoprotein (LDL) receptor (LDLR), and its deficiency in humans results in low plasma LDL cholesterol and protection against coronary heart disease. Recent evidence indicates that PCSK9 also modulates the metabolism of triglyceride-rich apolipoprotein B (apoB) lipoproteins, another important coronary heart disease risk factor. Here, we studied the effects of physiological levels of PCSK9 on intestinal triglyceride-rich apoB lipoprotein production and elucidated for the first time the cellular and molecular mechanisms involved. Treatment of human enterocytes (CaCo-2 cells) with recombinant human PCSK9 (10 μg/mL for 24 hours) increased cellular and secreted apoB48 and apoB100 by 40% to 55% each (P<0.01 versus untreated cells), whereas short-term deletion of PCSK9 expression reversed this effect. PCSK9 stimulation of apoB was due to a 1.5-fold increase in apoB mRNA (P<0.01) and to enhanced apoB protein stability through both LDLR-dependent and LDLR-independent mechanisms. PCSK9 decreased LDLR protein (P<0.01) and increased cellular apoB stability via activation of microsomal triglyceride transfer protein. PCSK9 also increased levels of the lipid-generating enzymes FAS, SCD, and DGAT2 (P<0.05). In mice, human PCSK9 at physiological levels increased intestinal microsomal triglyceride transfer protein levels and activity regardless of LDLR expression. PCSK9 markedly increases intestinal triglyceride-rich apoB production through mechanisms mediated in part by transcriptional effects on apoB, microsomal triglyceride transfer protein, and lipogenic genes and in part by posttranscriptional effects on the LDLR and microsomal triglyceride transfer protein. These findings indicate that targeted PCSK9-based therapies may also be effective in the management of postprandial hypertriglyceridemia.
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
DL-3-Hydroxy-3-methylglutaryl coenzyme A sodium salt hydrate, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK9
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcsk9
Supelco
Squalene, analytical standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Squalene, ≥98%, liquid