Merck
  • Probing the endocytic pathways of the filamentous bacteriophage in live cells using ratiometric pH fluorescent indicator.
Probing the endocytic pathways of the filamentous bacteriophage in live cells using ratiometric pH fluorescent indicator.
Advanced healthcare materials (2014-10-14)
Ye Tian, Man Wu, Xiangxiang Liu, Zhi Liu, Quan Zhou, Zhongwei Niu, Yong Huang
ABSTRACT
Viral nanoparticles have attracted extensive research interests in diverse applications of diagnosis and therapy. In particular, filamentous M13 bacteriophages have shown great potential in biomedical applications. However, its pathways entering into cells still remain unclear, and this greatly hinders its further use as a drug or gene carrier. Here, a ratiometric M13 pH probe is designed by conjugating two fluorescent dyes onto the surface of M13. Since the intensity ratio is not influenced by probe concentration, ion strength, temperature, photobleaching, and optical path length, this ratiometric probe can be used to investigate the intracellular pH map of M13. More importantly, the internalization mechanism of M13 can be elucidated. It is found that this filamentous phage shows great cell-type dependence in interaction with cells and internalization mechanism. The phage tends to be bounded on the cell membrane of only epithelial cells, not endothelial cells. Furthermore, the M13 phage enters into cells through endocytosis with specific mechanism: clathrin-mediated endocytosis and macropinocytosis for HeLa; vesicular transport, clathrin-mediated endocytosis, and macropinocytosis for MCF-7; caveolae-mediated endocytosis for human dermal microvascular endothelial cell (HDMEC). This work provides key notes for cancer diagnosis and therapy based on filamentous bacteriophage, especially for design of pH-sensitive drug delivery systems.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Supelco
Genistein, analytical standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers, BioReagent, suitable for protein labeling
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
USP
Amiloride hydrochloride, United States Pharmacopeia (USP) Reference Standard