Merck
  • Warfarin pretreatment reduces cell death and MMP-9 activity in experimental intracerebral hemorrhage.
Warfarin pretreatment reduces cell death and MMP-9 activity in experimental intracerebral hemorrhage.
Translational stroke research (2014-11-27)
Frieder Schlunk, Elena Schulz, Arne Lauer, Kazim Yigitkanli, Waltraud Pfeilschifter, Helmuth Steinmetz, Eng H Lo, Christian Foerch
ABSTRACT
Little is known about the pathophysiology of oral anticoagulation-associated intracerebral hemorrhage (OAC-ICH). We compared hematoma volume, number of terminal deoxynucleotidyl dUTP nick-end labeling (TUNEL)-positive cells (indicating cell death), MMP-9 levels, and perilesional edema formation between warfarin-treated mice and controls. Intracerebral hemorrhage was induced by an injection of collagenase into the right striatum. Twenty-four hours later, hematoma volume was measured using a photometric hemoglobin assay. Cell death was quantified using TUNEL staining. MMP-9 levels were determined by zymography, and edema formation was assessed via the wet-dry method. Warfarin increased hematoma volume by 2.6-fold. The absolute number of TUNEL-positive cells in the perihematomal zone was lower in warfarin-treated animals (300.5 ± 39.8 cells/mm2) than in controls (430.5 ± 38.9 cells/mm2; p = 0.034), despite the larger bleeding volume. MMP-9 levels were reduced in anticoagulated mice as compared to controls (p = 0.018). Perilesional edema formation was absent in warfarin mice and modestly present in controls. Our results suggest differences in the pathophysiology of OAC-ICH compared to intracerebral hemorrhage occurring under normal coagulation. A likely explanation is that thrombin, a strong inductor of apoptotic cell death and blood-brain barrier disruption, is produced to a lesser extent in OAC-ICH. In humans, however, we assume that the detrimental effects of a larger hematoma volume in OAC-ICH by far outweigh potential protective effects of thrombin deficiency.
Product Description
Supelco
Warfarin, analytical standard
Warfarin sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Warfarin Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Warfarin, PESTANAL®, analytical standard
USP
Warfarin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, ≥98%