Merck
  • Assessing positivity and circulating levels of NS1 in samples from a 2012 dengue outbreak in Rio de Janeiro, Brazil.
Assessing positivity and circulating levels of NS1 in samples from a 2012 dengue outbreak in Rio de Janeiro, Brazil.
PloS one (2014-11-21)
Diego Allonso, Marcelo D F Meneses, Carlos A Fernandes, Davis F Ferreira, Ronaldo Mohana-Borges
ABSTRACT
Dengue virus (DENV) represents a major threat to public health worldwide. Early DENV diagnosis should not only detect the infection but also identify patients with a higher likelihood to develop severe cases. Previous studies have suggested the potential for NS1 to serve as a viral marker for dengue severity. However, further studies using different sera panels are required to confirm this hypothesis. In this context, we developed a lab-based ELISA to detect and quantitate NS1 protein from the four DENV serotypes and from primary and secondary cases. This approach was used to calculate the circulating NS1 concentration in positive samples. We also tested the NS1 positivity of DENV-positive samples according to the Platelia Dengue NS1 Ag assay. A total of 128 samples were positive for DENV infection and were classified according to the WHO guidelines. The overall NS1 positivity was 68% according to the Platelia assay, whereas all samples were NS1-positive when analyzed with our lab-based ELISA. Fifty-four samples were positive by PCR, revealing a co-circulation of DENV1 and DENV4, and the NS1 positivity for DENV4 samples was lower than that for DENV1. The circulating NS1 concentration ranged from 7 to 284 ng/mL. Our results support previous data indicating the low efficiency of the Platelia assay to detect DENV4 infection. Moreover, this work is the first to analyze NS1 antigenemia using retrospective samples from a Brazilian outbreak.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, ≥99%, AR grade
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, AR, ≥99.9%
Sigma-Aldrich
Anti-NS1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate, 96%