Merck
  • Characterizing the impact of traffic and the built environment on near-road ultrafine particle and black carbon concentrations.
Characterizing the impact of traffic and the built environment on near-road ultrafine particle and black carbon concentrations.
Environmental research (2014-05-20)
Scott Weichenthal, William Farrell, Mark Goldberg, Lawrence Joseph, Marianne Hatzopoulou
ABSTRACT
Increasing evidence suggests that ultrafine particles (UFPs) may contribute to cardiorespiratory morbidity. We examined the relationship between near road UFPs and several traffic and built environment factors to identify predictors that may be used to estimate exposures in population-based studies. Black carbon (BC) was also examined. Data were collected on up to 6 occasions at 73 sites in Montreal, Canada over 6-week period during summer, 2012. After excluding highly correlated variables, road width, truck ratio (trucks/total traffic), building height, land zoning parameters, and meteorological factors were evaluated. Random-effect models were used to estimate percent changes in UFP and BC concentrations with interquartile changes in each candidate predictor adjusted for meteorological factors. Mean pollutant concentrations varied substantially across sites (UFP range: 1977-94, 798 particles/cm(3); BC range: 29-9460 ng/m(3)). After adjusting for meteorology, interquartile increases in road width (14%, 95% CI: 0, 30), building height (13%, 95% CI: 5, 22), and truck ratio (13%, 95% CI: 3, 23) were the most important predictors of mean UFP concentrations. Road width (28%, 95% CI: 9, 51) and industrial zoning (18%, 95% CI: 2, 37) were the strongest predictors of maximum UFP concentrations. Industrial zoning (35%, 95% CI: 9, 67) was the strongest predictor of BC. A number of traffic and built environmental factors were identified as important predictors of near road UFP and BC concentrations. Exposure models incorporating these factors may be useful in evaluating the health effects of traffic related air pollution.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Millipore
Activated charcoal, suitable for GC
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%