Merck
  • Myoclonus and epilepsy in childhood: a review of treatment with valproate, ethosuximide, lamotrigine and zonisamide.
Myoclonus and epilepsy in childhood: a review of treatment with valproate, ethosuximide, lamotrigine and zonisamide.
Epilepsy research (1998-02-26)
S J Wallace
ABSTRACT
Based on small numbers of patients, it is possible to make the following suggestions rather than categorical statements. For myoclonic seizures and epilepsies which are not otherwise specified, valproate seems of proven efficacy. Ethosuximide may be a useful adjunct. The exact place of lamotrigine, which controls some myoclonia and makes them worse in other patients, requires further study. The findings are clearer when specific syndromes are considered. Valproate is the treatment of first choice for benign myoclonic epilepsy in infants, myoclonic astatic epilepsy, epilepsy with myoclonic absences, eyelid myoclonia with absences, juvenile myoclonic epilepsy and progressive myoclonus epilepsy. The addition of ethosuximide to valproate can be helpful to those with myoclonic absences, where this combination appears more beneficial than either valproate or ethosuximide alone and in eyelid myoclonia with absences. Lamotrigine can be effective therapy for juvenile myoclonic epilepsy and eyelid myoclonia with absences when used alone and, in conjunction with other antiepileptic drugs (AED) (usually valproate) for early myoclonic encephalopathy, myoclonic-astatic epilepsy and particularly, epilepsy with myoclonic absences. The myoclonia of infantile neuronal ceroid lipofuscinosis respond to lamotrigine. Severe myoclonic epilepsy of infants usually worsens with lamotrigine, but occasionally, children improve. Zonisamide added to clonazepam and valproate or a barbiturate, can reduce the cascade of myoclonia in progressive myoclonus epilepsies for at least 2 years, but relapse may occur thereafter.
Sigma-Aldrich
Ethosuximide
Supelco
Ethosuximide, Pharmaceutical Secondary Standard; Certified Reference Material