Merck
  • A meta-analytic review of ERCC1/MDR1 polymorphism and chemosensitivity to platinum in patients with advanced non-small cell lung cancer.
A meta-analytic review of ERCC1/MDR1 polymorphism and chemosensitivity to platinum in patients with advanced non-small cell lung cancer.
Chinese medical journal (2012-08-31)
Hai-Bo Wei, Jing Hu, Li-Hua Shang, Yun-Yan Zhang, Fei-Fei Lu, Min Wei, Yan Yu
ABSTRACT
Platinum-based regimens are used as standard first-line chemotherapy in non-small cell lung cancer (NSCLC) patients. To study if pharmacogenetic approach may allow a tailored selection of platinum chemotherapy for advanced NSCLC, we performed a meta-analysis to compare chemosensitivity to platinum with different ERCC1 C118T/ MDR1 C3435T single-nucleotide polymorphism (SNP). Relevant studies were identified by searching the PubMed, Embase, Cochrane, OVID, Springer, EBSCO and CNKI databases. Inclusion criteria were patients with advanced NSCLC who received platinum-based chemotherapy, an evaluated polymorphism of ERCC/MDR1 and overall response rate. We excluded duplicate publications, letters and review articles. The RevMan 4.2 and STATA 11 package were used to do comprehensive quantitative assessment. A total of 11 studies were included in this meta-analysis. For studies evaluating ERCC1 C118T, test for heterogeneity was done (χ(2) = 13.41, P = 0.1), and the odds ratio (OR) for the wild-type C/C genotype versus the heterozygous C/T and T/T genotypes was 1.50 (95% CI 1.09 - 2.06, P = 0.01). In four studies evaluating MDR1 polymorphism, test for heterogeneity was also done (χ(2) = 3.22, P = 0.36), and the OR for the wild-type C/C genotype versus the heterozygous C/T and T/T genotypes was 2.30 (95% CI 1.44 - 3.68, P = 0.0005). The results indicated that platinum-based chemotherapy sensitivity was significantly associated with polymorphism of ERCC1 C118T and MDR1 C3435T SNP. In further perspective studies, the ERCC1/MDR1 SNPs might serve as simple and less invasive biomarkers for personalized chemotherapy with platinum-based anticancer drugs.
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Platinum, foil, 0.2m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Sigma-Aldrich
Platinum, nanopowder, 200 nm particle size (SEM), 99.9% (metals basis)
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%
Platinum, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.998%
Platinum, wire reel, 0.1m, diameter 0.15mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, wire reel, 0.1m, diameter 0.006mm, wollaston, 99.9%
Platinum, foil, 0.1m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.003mm, as rolled, 99.95%
Platinum, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.001mm, PTFE (polytetrafluoroethylene) insulation, 99.999%
Platinum, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99.95%
Platinum, tube, 100mm, outside diameter 0.7mm, inside diameter 0.4mm, wall thickness 0.15mm, as drawn, 99.95%
Platinum, wire reel, 0.05m, diameter 0.8mm, annealed, may be used as one component of a thermocouple, 99.99%
Platinum, rod, 25mm, diameter 1.0mm, 99.95%