Merck
  • Hydroquinone therapy for post-inflammatory hyperpigmentation secondary to acne: not just prescribable by dermatologists.
Hydroquinone therapy for post-inflammatory hyperpigmentation secondary to acne: not just prescribable by dermatologists.
Acta dermato-venereologica (2011-10-18)
Mekhala Chandra, Jacob Levitt, Cara A Pensabene
ABSTRACT
Post-inflammatory hyperpigmentation after acne can be as troublesome as the acne itself. Hydroquinone, a tyrosinase inhibitor, in a 4% cream can be used safely twice daily for up to 6 months to treat post-inflammatory hyperpigmentation. The efficacy of this treatment can be enhanced by using a retinoid nightly and a mid-potent steroid, which is applied twice daily for 2 weeks, then at weekends only. Combination creams help with compliance, but often lack the strongest individual ingredients. Because steroids should not be applied to the face for prolonged periods, care should be taken when a hydroquinone cream containing a steroid is chosen. If post-inflammatory hyperpigmentation consists of a few lesions, spot therapy is useful. If post-inflammatory hyperpigmentation consists of many lesions, field therapy is favored. Safety concerns with hydroquinone consist only of occasional irritation, which can be suppressed with topical steroid or a short drug holiday. Physicians should feel comfortable to use hydroquinone without consulting a dermatologist. Key words: hydroquinone; acne; adolescent; post-inflammatory hyperpigmentation.
Product Description
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland