Merck
Leaching of plastic additives to marine organisms.
Environmental pollution (Barking, Essex : 1987) (2014-01-21)
Albert A Koelmans, Ellen Besseling, Edwin M Foekema
ABSTRACT
It is often assumed that ingestion of microplastics by aquatic species leads to increased exposure to plastic additives. However, experimental data or model based evidence is lacking. Here we assess the potential of leaching of nonylphenol (NP) and bisphenol A (BPA) in the intestinal tracts of Arenicola marina (lugworm) and Gadus morhua (North Sea cod). We use a biodynamic model that allows calculations of the relative contribution of plastic ingestion to total exposure of aquatic species to chemicals residing in the ingested plastic. Uncertainty in the most crucial parameters is accounted for by probabilistic modeling. Our conservative analysis shows that plastic ingestion by the lugworm yields NP and BPA concentrations that stay below the lower ends of global NP and BPA concentration ranges, and therefore are not likely to constitute a relevant exposure pathway. For cod, plastic ingestion appears to be a negligible pathway for exposure to NP and BPA.
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, ≥99%
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland