Merck
  • ZnO nanostructure construction on zinc foil: the concept from an ionic liquid precursor aqueous solution.
ZnO nanostructure construction on zinc foil: the concept from an ionic liquid precursor aqueous solution.
Chemical communications (Cambridge, England) (2009-10-15)
Zhonghao Li, Yuxia Luan, Qingzhong Wang, Guangshan Zhuang, Yongxin Qi, Yuan Wang, Chengguo Wang
ABSTRACT
Aligned ZnO arrays with branched rods, hexagonal rods, quasi-round rods, hexagonal pyramids and porous nanostructures are constructed on zinc foil in the ionic liquid tetrabutylammonium hydroxide aqueous solution.
Product Description
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 4mm disks, thickness 0.15mm, as rolled, 99.95+%