Merck
  • Detection of amitriptyline, nortriptyline and bromazepam in liver, CSF and hair in the homicidal poisoning of a one-month-old girl autopsied 8 months after death.
Detection of amitriptyline, nortriptyline and bromazepam in liver, CSF and hair in the homicidal poisoning of a one-month-old girl autopsied 8 months after death.
Forensic science international (2011-01-05)
Yvan Gaillard, Rémi Breuil, Christophe Doche, Ludovic Romeuf, Catherine Lemeur, Jean Michel Prevosto, Laurent Fanton
ABSTRACT
We reported on the death by poisoning of a one-month-old baby that had followed the death of one of her sister (due to cyamemazine overdose). Exhumation of the corpse was done 8 months after burial and revealed the presence of amitriptyline. Parent drug and its metabolite were analysed by HPLC-MS/MS in positive ionisation mode on a C(18) analytical column using a gradient of acetonitrile and 2mM formate buffer at pH=3. Quantification is based on the main ion m/z=233, the common product ion of nortriptyline (MH(+), m/z 264), amitriptyline (MH(+), m/z 278) and nortriptyline D3 used as internal standard (MH(+), m/z 267). Amitriptyline and nortriptyline in the liver were measured at a concentration of 29.8 and 3.6 μg/g, respectively. Hair analyses revealed the presence of amitriptyline and nortriptyline at concentrations of 1811 and 43 pg/mg, respectively, while complementary analyses showed the presence of bromazepam in the hair at a concentration of 740 pg/mg, thus documenting previous administrations. The mother confessed later having used the drinkable form of the pharmaceutical LAROXYL(®) by pouring the content of a 20 ml bottle (at 40 mg/ml) into the feeding-bottle of her child. The milk was sweet but still bitter and following the testimony of a close relative, the whole family helped to feed the crying baby.
Product Description
Supelco
Bromazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®