Merck
  • FD&C Red No. 105 (Rose Bengal B) neutralizes the thyroid tumor promoting effect of iodine-deficient diet in rats.
FD&C Red No. 105 (Rose Bengal B) neutralizes the thyroid tumor promoting effect of iodine-deficient diet in rats.
Toxicology (1995-05-05)
J Kanno, Y Goda, K Sato, K Yoshihira, Y Hayashi
ABSTRACT
The effect of 3,4,5,6-tetrachloro-2',4',5',7'-tetraiodo-fluorescein sodium salt (Rose Bengal B or FD&C Red No. 105, molecular weight 1017.6) on the thyroid of rats treated with N-bis(2-hydroxypropyl)-nitrosamine (DHPN) and iodine-deficient (I-def) diet is studied. Six-week-old male F344 rats were divided into seven groups (20 each) and given a single subcutaneous injection of DHPN (2800 mg/kg body wt.). From week 2 to 20, I-def diet was given in combination with either FR105 (1.25, 5.0, or 20 mg/l) or potassium iodide (KI, 12.5, 50.0 or 200 micrograms/l) in drinking water. As a result, an amount of 1.25 mg/l of FR105 was slightly more effective than 200 micrograms/l of KI in terms of inhibition of the effect of I-def diet on thyroid weight, morphology, thyroid-related hormones and thyroid tumor development. It was calculated that 1 mumol/l of FR105 was slightly more potent than 1 mumol/l of iodide ion. As each FR105 molecule has four iodide residues, at least 25% of total iodide residues were calculated to be utilized by the rats given I-def diet.
Sigma-Aldrich
Rose bengal, certified by the BSC, Dye content ≥80 %
Sigma-Aldrich
Rose Bengal lactone, 95%
Sigma-Aldrich
Rose bengal, Dye content 95 %, ≥96% (HPLC)