Merck
  • Degradation of triphenylmethane dyes using a temperature and pH stable spore laccase from a novel strain of Bacillus vallismortis.
Degradation of triphenylmethane dyes using a temperature and pH stable spore laccase from a novel strain of Bacillus vallismortis.
Bioresource technology (2012-10-18)
Chong Zhang, Hanwen Diao, Fengxia Lu, Xiaomei Bie, Yufeng Wang, Zhaoxin Lu
ABSTRACT
The characterization of a spore laccase from Bacillus vallismortis fmb-103, isolated from textile industry disposal sites, is described. The activity was 6.5 U/g of dry spore with ABTS as the substrate. The enzyme was quite stable at high temperature. It retained more than 90% of its initial activity after 10h at 70 °C. The enzyme demonstrated broad pH stability in both acidic and alkaline conditions. There was almost no activity loss at pH 3 over an extended period of time, and the relative activity remained at 82% and 38% at pH 7 and pH 9 after 10 days. NaN(3), SDS, L-cysterine, Dithiothreitol, EDTA and NaCl inhibit the enzyme activity. Triphenylmethane dyes, including malachite green, brilliant green and aniline blue were efficiently degraded by the enzyme after 24h in combination with a mediator with efficiencies of 76.84%, 96.56% and 81.17%, respectively. The reusability of spore laccase for decolorization dyes was also examined.
Sigma-Aldrich
Triphenylmethane, 99%