Merck
  • Colonoscopy is superior to neostigmine in the treatment of Ogilvie's syndrome.
Colonoscopy is superior to neostigmine in the treatment of Ogilvie's syndrome.
American journal of surgery (2012-10-02)
Victor B Tsirline, Alla Y Zemlyak, Michael J Avery, Paul D Colavita, Ashley B Christmas, B Todd Heniford, Ronald F Sing
ABSTRACT
Colonic pseudo-obstruction in critically ill patients may lead to devastating colonic perforation. Neostigmine is often the first-line intervention, because colonoscopy is more invasive and labor intensive. A retrospective 10-year review at a tertiary medical center identified 100 patients with Ogilvie's syndrome, in whom treatment course and clinical and radiographic response were evaluated. Colonoscopy was significantly more successful than neostigmine (defined as no further therapy) after 1 or 2 interventions (75.0% vs 35.5%, P = .0002, and 84.6% vs 55.6%, P = .0031, respectively). One colonoscopy was more effective than 2 neostigmine administrations (75.0% vs 55.6%, P = .044). Clinical response (poor, fair, or good) was significantly better after colonoscopy than neostigmine after 1 or 2 interventions (P = .0028 and P = .00079). Cecal diameters decreased significantly more after colonoscopy than neostigmine (from 10.2 ± .5 cm to 7.1 ± .4 cm vs from 10.5 ± .5 cm to 8.8 ± .5 cm, P = .026). Neostigmine administration before colonoscopy did not affect outcomes. There were 3 perforations (3.7%): 1 each after colonoscopy, neostigmine, and no intervention. Neostigmine dose or repetition did not affect radiographic (P = .41) or clinical (P = .31) response. Colonoscopy is superior to neostigmine for Ogilvie's syndrome and should be considered first-line therapy, although neostigmine is useful in select patients and repeat interventions.
Sigma-Aldrich
Neostigmine bromide, ≥98% (HPLC and titration), powder
Sigma-Aldrich
Neostigmine methyl sulfate
Neostigmine methyl sulfate, European Pharmacopoeia (EP) Reference Standard