Merck
  • Vibrational spectra and density functional theoretical calculations on the anti-neurodegenerative drug: Orphenadrine hydrochloride.
Vibrational spectra and density functional theoretical calculations on the anti-neurodegenerative drug: Orphenadrine hydrochloride.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-21)
Bismi Edwin, I Hubert Joe
ABSTRACT
Vibrational spectral analysis and quantum chemical computations based on density functional theory have been performed on the anti-neuro-degenerative drug Orphenadrine hydrochloride. The geometry, intermolecular hydrogen bond, and harmonic vibrational frequencies of the title molecule have been investigated with the help of B3LYP method. The calculated molecular geometry has been compared with the experimental data. The various intramolecular interactions have been exposed by natural bond orbital analysis. The distribution of Mulliken atomic charges and bending of natural hybrid orbitals also reflect the presence of intramolecular hydrogen bonding. The analysis of the electron density of HOMO and LUMO gives an idea of the delocalization and low value of energy gap indicates electron transport in the molecule and thereby bioactivity. Effective docking of the drug molecule with NMDA receptor subunit 3A also enhances its bioactive nature.
Product Description
Supelco
Orphenadrine citrate salt, analytical standard
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)