Merck
  • Chemical functionalization of bioceramics to enhance endothelial cells adhesion for tissue engineering.
Chemical functionalization of bioceramics to enhance endothelial cells adhesion for tissue engineering.
Journal of medicinal chemistry (2012-08-18)
Françoise Borcard, Davide Staedler, Horacio Comas, Franziska Krauss Juillerat, Philip N Sturzenegger, Roman Heuberger, Urs T Gonzenbach, Lucienne Juillerat-Jeanneret, Sandrine Gerber-Lemaire
ABSTRACT
To control the selective adhesion of human endothelial cells and human serum proteins to bioceramics of different compositions, a multifunctional ligand containing a cyclic arginine-glycine-aspartate (RGD) peptide, a tetraethylene glycol spacer, and a gallate moiety was designed, synthesized, and characterized. The binding of this ligand to alumina-based, hydroxyapatite-based, and calcium phosphate-based bioceramics was demonstrated. The conjugation of this ligand to the bioceramics induced a decrease in the nonselective and integrin-selective binding of human serum proteins, whereas the binding and adhesion of human endothelial cells was enhanced, dependent on the particular bioceramics.
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)