Merck
  • Efficacy and safety of aceclofenac-cr and aceclofenac in the treatment of knee osteoarthritis: a 6-week, comparative, randomized, multicentric, double-blind study.
Efficacy and safety of aceclofenac-cr and aceclofenac in the treatment of knee osteoarthritis: a 6-week, comparative, randomized, multicentric, double-blind study.
The journal of pain : official journal of the American Pain Society (2011-02-01)
Anil Pareek, Nitin Chandurkar, Anil Gupta, Ashish Sirsikar, Bhavik Dalal, Bhavesh Jesalpura, Anoop Mehrotra, Arunangshu Mukherjee
ABSTRACT
The efficacy and safety of aceclofenac control release (CR) tablets was compared with conventional aceclofenac tablets in patients with knee osteoarthritis (OA). This was a double-blind, double-dummy, randomized, parallel group multicentric study conducted at 6 centers. Two hundred and eighty five patients were randomized to either aceclofenac-CR (n = 143) once daily or conventional aceclofenac tablet (n = 142) twice daily and were followed for 6 weeks. The efficacy parameters were pain intensity score on visual analogue scale, Western Ontario and McMaster (WOMAC) score, patients and investigator's overall study drug assessment and total consumption of acetaminophen and ranitidine tablets. Both treatments showed significant improvement in their efficacy parameters from baseline at the end of therapy. Aceclofenac-CR was comparable to conventional aceclofenac with respect to change in pain intensity and WOMAC score (P > .05) There was no statistically significant difference between the treatment groups in patient's and investigator's overall study drug assessment at the end of therapy (P > .05). Aceclofenac-CR treated patients took fewer acetaminophen and ranitidine tablets during the treatment period as compared to conventional aceclofenac treated patients. Both the study medications were well tolerated with no incidence of serious adverse event (SAE). In conclusion, the new aceclofenac-CR formulation was found to be effective and safe while offering practical advantage of once daily administration. This article represents the advantages of control release aceclofenac over the conventional aceclofenac tablets. Aceclofenac-CR was found to be similar in terms of efficacy as conventional aceclofenac in knee OA patients with fewer adverse events.
Product Description
Aceclofenac for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aceclofenac, ≥98% (HPLC)