Merck
  • Self-assembly and characterization of Pluronic P105 micelles for liver-targeted delivery of silybin.
Self-assembly and characterization of Pluronic P105 micelles for liver-targeted delivery of silybin.
Journal of drug targeting (2009-10-30)
Xinru Li, Yanqing Huang, Xingwei Chen, Yanxia Zhou, Yanhui Zhang, Pingzhu Li, Yan Liu, Yufeng Sun, Jieyu Zhao, Fei Wang
ABSTRACT
Polymeric micelles, based on lactobionic acid (LA)-conjugated Pluronic P105 (P105), were prepared to achieve liver-targeted delivery of silybin. In the triblock copolymer structure of PEO-PPO-PEO, LA was successfully conjugated with the terminal end of PEO to produce LA-P105. The success of synthesis was confirmed using FTIR and 1H NMR. The triblock copolymers with functional moiety were physically mixed with silybin to form micelles. Silybin-loaded LA-P105 micelles characterized by dynamic light scattering and transmission electron microscopy (TEM) were uniform spherical particles. There was a remarkable increase in the dissolubility for silybin in LA-P105 micelle solution (627 microg/mL) when compared with that for water (4.6 microg/mL). The pharmacokinetic experiments showed that the area under the curve of silybin plasma concentration-time profile in rats for LA-P105 micelles was lower than that for P105 micelles. Biodistribution studies indicated that a significantly increased amount of silybin was accumulated in liver, suggesting that LA locating on the surface of the micelles played an important role in transporting an increased amount of silybin into liver. This polymeric vehicle is, therefore, expected to be widely used as target-specific delivery vehicles for diverse water-insoluble therapeutic and diagnostic agents.
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Lactobionic acid, European Pharmacopoeia (EP) Reference Standard