Merck
  • Reactivity to sodium tetrachloropalladate (Na2[PdCl4]) compared to PdCl2 and NiCl2 in lymphocyte proliferation tests.
Reactivity to sodium tetrachloropalladate (Na2[PdCl4]) compared to PdCl2 and NiCl2 in lymphocyte proliferation tests.
Allergy (2009-02-12)
J Muris, C J Kleverlaan, A J Feilzer, E Valentine-Thon
ABSTRACT
For patch testing, replacement of the commonly used palladium dichloride (PdCl2) by sodium tetrachloropalladate (Na2[PdCl4]) was recently demonstrated to improve test accuracy and show a significant correlation with nickel (Ni), supporting the concept of cross-reactivity between Pd and Ni. A promising alternative to metal allergy patch testing is the in vitro lymphocyte proliferation test (LTT). The aim of this study was to test whether Na2[PdCl4] is also more sensitive for diagnosing Pd allergy with a standardized LTT. After determining optimal nontoxic and nonmitogenic concentrations for Na2[PdCl4], blood samples from 105 patients with clinical suspicion of metal allergy were tested with an LTT called memory lymphocyte immuno stimulation assay for Na2[PdCl4], PdCl2 and NiCl2. Reaction profiles were analysed for concordant positive reactions. Using the conventional cut-off of stimulation index > or = 3, 74.3% showed a positive reaction to NiCl2, 15.2% to PdCl2 and 28.6% to Na2[PdCl4]. All positive results to PdCl2 were covered by Na2[PdCl4]. From the 30 positive reactions to Na2[PdCl4], 26 (87%) were concordant for NiCl2 reactivity. In LTT, the use of Na2[PdCl4] results in more positive reactions in Pd allergy testing which are in concordance with positive reactions to PdCl2 and NiCl2.
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis