Merck
  • Control of protein structure and function through surface recognition by tailored nanoparticle scaffolds.
Control of protein structure and function through surface recognition by tailored nanoparticle scaffolds.
Journal of the American Chemical Society (2004-01-22)
Rui Hong, Nicholas O Fischer, Ayush Verma, Catherine M Goodman, Todd Emrick, Vincent M Rotello
ABSTRACT
Thioalkyl and thioalkylated oligo(ethylene glycol) (OEG) ligands with chain-end functionality were used to fabricate water-soluble CdSe nanoparticle scaffolds. Surface recognition of chymotrypsin (ChT) was achieved using these functionalized nanoparticle scaffolds, with three levels of interaction demonstrated: no interaction (OEG terminated with hydroxyl group), inhibition with denaturation (carboxylate-terminated thioalkyl ligands), and inhibition with retention of structure (carboxylate-terminated OEG). The latter process was reversible upon an increase in ionic strength, with essentially complete restoration of enzymatic activity.
Sigma-Aldrich
Undecanoic acid, 98%
Supelco
Undecanoic acid, analytical standard
Sigma-Aldrich
Undecanoic acid, ≥99%, FG