Merck
  • Leishmania (Viannia) braziliensis nucleoside triphosphate diphosphohydrolase (NTPDase 1): localization and in vitro inhibition of promastigotes growth by polyclonal antibodies.
Leishmania (Viannia) braziliensis nucleoside triphosphate diphosphohydrolase (NTPDase 1): localization and in vitro inhibition of promastigotes growth by polyclonal antibodies.
Experimental parasitology (2012-08-28)
Gabriane Nascimento Porcino, Cristiane Carvalho-Campos, Ana Carolina Ribeiro Gomes Maia, Michelle Lima Detoni, Priscila Faria-Pinto, Elaine Soares Coimbra, Marcos José Marques, Maria Aparecida Juliano, Luiz Juliano, Vanessa Álvaro Diniz, Suzana Corte-Real, Eveline Gomes Vasconcelos
ABSTRACT
Nucleoside triphosphate diphosphohydrolase (NTPDase) activity was recently characterized in Leishmania (Viannia) braziliensis promastigotes (Lb), and an antigenic conserved domain (r82-121) from the specific NTPDase 1 isoform was identified. In this work, mouse polyclonal antibodies produced against two synthetic peptides derived from this domain (LbB1LJ, r82-103; LbB2LJ, r102-121) were used. The anti-LbB1LJ or anti-LbB2LJ antibodies were immobilized on protein A-sepharose and immunoprecipitated the NTPDase 1 of 48 kDa and depleted approximately 40% of the phosphohydrolytic activity from detergent-homogenized Lb preparation. Ultrastructural immunocytochemical microscopy identified the NTPDase 1 on the parasite surface and in its subcellular cytoplasmic vesicles, mitochondria, kinetoplast and nucleus. The ATPase and ADPase activities of detergent-homogenized Lb preparation were partially inhibited by anti-LbB1LJ antibody (43-79%), which was more effective than that inhibition (18-47%) by anti-LbB2LJ antibody. In addition, the immune serum anti-LbB1LJ (67%) or anti-LbB2LJ (33%) was cytotoxic, significantly reducing the promastigotes growth in vitro. The results appoint the conserved domain from the L. braziliensis NTPDase as an important target for inhibitor design and the potential application of these biomolecules in experimental protocols of disease control.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder