Merck
  • Serosurveillance for Japanese encephalitis, Akabane, and Aino viruses for Thoroughbred horses in Korea.
Serosurveillance for Japanese encephalitis, Akabane, and Aino viruses for Thoroughbred horses in Korea.
Journal of veterinary science (2008-12-02)
Dong Kun Yang, Byoung Han Kim, Chang Hee Kweon, Jin Ju Nah, Hyun Joo Kim, Kyung Woo Lee, Young Jin Yang, Kyu Whan Mun
ABSTRACT
Recent global warming trends may have a significant impact on vector-borne viral diseases, possibly affecting vector population dynamics and disease transmission. This study measured levels of hemagglutination-inhibition (HI) antibodies against Japanese encephalitis virus (JEV) and neutralizing antibodies against Akabane virus (AKAV) and Aino virus (AINV) for Thoroughbred horses in Korea. Blood samples were collected from 989 racehorses in several provinces, between October 2005 and March 2007. Sera were tested using either an HI assay or a virus neutralization test. Approximately half (49.7%; 492/989) of the horses tested were antibody-positive for JEV. The HI titer against JEV was significantly correlated with racehorse age (p < 0.05). Horses with an HI antibody titer of 1 : 160 or higher accounted for 3.9% of the animals tested, indicating that vectors transmitting arthropod- borne viruses bit relatively few horses. In contrast, 3.8% (19/497) and 19.5% (97/497) of horse sera collected in March 2007 were positive against AKAV and AINV, respectively. The presence of antibodies against AKAV and AINV may indicate the multiplication of AKAV and AINV in these horses.
Sigma-Aldrich
Kaolin
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin, meets USP testing specifications