Merck
  • Preparation of the membrane-permeant biarsenicals FlAsH-EDT2 and ReAsH-EDT2 for fluorescent labeling of tetracysteine-tagged proteins.
Preparation of the membrane-permeant biarsenicals FlAsH-EDT2 and ReAsH-EDT2 for fluorescent labeling of tetracysteine-tagged proteins.
Nature protocols (2008-09-06)
Stephen R Adams, Roger Y Tsien
ABSTRACT
The membrane-permeant fluorogenic biarsenicals FlAsH-EDT(2) and ReAsH-EDT(2) can be prepared in good yields by a straightforward two-step procedure from the inexpensive precursor dyes fluorescein and resorufin, respectively. Handling of toxic reagents such as arsenic trichloride is minimized so the synthesis can be carried out in a typical chemistry laboratory, usually taking about 2-3 d. A wide range of other biarsenical reagents and intermediates that also bind to tetracysteine-tagged (CysCysProGlyCysCys) proteins can be prepared similarly using this general procedure.
Product Description
Supelco
Sodium 2-mercaptoethanesulfonate, analytical standard, ≥98.0% (titration)
Sigma-Aldrich
Aldrich® Firestone valve, controls gas flow and pressure in reaction vessels
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Palladium(II) acetate, reagent grade, 98%
Sigma-Aldrich
Palladium(II) Acetate ChemBeads
Sigma-Aldrich
1,2-Ethanedithiol, technical grade, ≥90%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%