Merck
  • Chronic kidney disease may evoke anxiety by altering CRH expression in the amygdala and tryptophan metabolism in rats.
Chronic kidney disease may evoke anxiety by altering CRH expression in the amygdala and tryptophan metabolism in rats.
Pflugers Archiv : European journal of physiology (2023-11-22)
Katalin Eszter Ibos, Éva Bodnár, Hoa Dinh, Merse Kis, Fanni Márványkövi, Zsuzsanna Z A Kovács, Andrea Siska, Imre Földesi, Zsolt Galla, Péter Monostori, István Szatmári, Péter Simon, Márta Sárközy, Krisztina Csabafi
ABSTRACT
Chronic kidney disease (CKD) is associated with anxiety; however, its exact mechanism is not well understood. Therefore, the aim of the present study was to assess the effect of moderate CKD on anxiety in rats. 5/6 nephrectomy was performed in male Wistar rats. 7 weeks after, anxiety-like behavior was assessed by elevated plus maze (EPM), open field (OF), and marble burying (MB) tests. At weeks 8 and 9, urinalysis was performed, and blood and amygdala samples were collected, respectively. In the amygdala, the gene expression of Avp and the gene and protein expression of Crh, Crhr1, and Crhr2 were analyzed. Furthermore, the plasma concentration of corticosterone, uremic toxins, and tryptophan metabolites was measured by UHPLC-MS/MS. Laboratory tests confirmed the development of CKD. In the CKD group, the closed arm time increased; the central time and the total number of entries decreased in the EPM. There was a reduction in rearing, central distance and time in the OF, and fewer interactions with marbles were detected during MB. CKD evoked an upregulation of gene expression of Crh, Crhr1, and Crhr2, but not Avp, in the amygdala. However, there was no alteration in protein expression. In the CKD group, plasma concentrations of p-cresyl-sulfate, indoxyl-sulfate, kynurenine, kynurenic acid, 3-hydroxykynurenine, anthranilic acid, xanthurenic acid, 5-hydroxyindoleacetic acid, picolinic acid, and quinolinic acid increased. However, the levels of tryptophan, tryptamine, 5-hydroxytryptophan, serotonin, and tyrosine decreased. In conclusion, moderate CKD evoked anxiety-like behavior that might be mediated by the accumulation of uremic toxins and metabolites of the kynurenine pathway, but the contribution of the amygdalar CRH system to the development of anxiety seems to be negligible at this stage.
Sigma-Aldrich
Anti-CRHR1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CRHR2 antibody produced in rabbit, affinity isolated antibody