Merck
  • Prolactin enhances T regulatory cell promotion of breast cancer through the long form prolactin receptor.
Prolactin enhances T regulatory cell promotion of breast cancer through the long form prolactin receptor.
Translational oncology (2021-08-11)
Kuan-Hui Ethan Chen, Mrinal Ghosh, Lorena Rivera, Samuel Lin, Anil Kumar, Srividya Swaminathan, Mary Y Lorenson, Ameae M Walker
ABSTRACT
Previous work has shown systemic knockdown of the long form prolactin receptor (LFPRLR) in vivo markedly reduced metastasis in mouse models of breast cancer, but whether this translated to prolonged survival was unknown. Here we show that LFPRLR knockdown in the highly metastatic, immunocompetent 4T1 model prolonged survival and reduced recruitment of T regulatory cells (Tregs) to the tumor through effects on the production of CCL17. For the Tregs still recruited to the primary tumor, LFPRLR knockdown both directly and indirectly reduced their ability to promote tumor parenchymal epithelial to mesenchymal transition. Importantly, effects of prolactin on expression of mesenchymal genes by the tumor parenchyma were very different in the absence and presence of Tregs. While systemic knockdown of the LFPRLR downregulated transcripts important for immune synapse function in the remaining tumor Tregs, splenic Tregs seemed unaffected by LFPRLR knockdown, as demonstrated by their continued ability to suppress anti-CD3/CD28-stimulated effector cell proliferation at 1-5 months. These results demonstrate that knockdown of the LFPRLR achieves intra-tumor immunotherapeutic effects and suggest this occurs with reduced likelihood of peripheral inflammatory/autoimmune sequelae.
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture