Merck
  • Deletion at an 1q24 locus reveals a critical role of long noncoding RNA DNM3OS in skeletal development.
Deletion at an 1q24 locus reveals a critical role of long noncoding RNA DNM3OS in skeletal development.
Cell & bioscience (2021-03-04)
Ting-Ting Yu, Qiu-Fan Xu, Si-Yang Li, Hui-Jie Huang, Sarah Dugan, Lei Shao, Jennifer A Roggenbuck, Xiao-Tong Liu, Huai-Ze Liu, Betsy A Hirsch, Shen Yue, Chen Liu, Steven Y Cheng
ABSTRACT
Skeletal development and maintenance are complex processes known to be coordinated by multiple genetic and epigenetic signaling pathways. However, the role of long non-coding RNAs (lncRNAs), a class of crucial epigenetic regulatory molecules, has been under explored in skeletal biology. Here we report a young patient with short stature, hypothalamic dysfunction and mild macrocephaly, who carries a maternally inherited 690 kb deletion at Chr.1q24.2 encompassing a noncoding RNA gene, DNM3OS, embedded on the opposite strand in an intron of the DYNAMIN 3 (DNM3) gene. We show that lncRNA DNM3OS sustains the proliferation of chondrocytes independent of two co-cistronic microRNAs miR-199a and miR-214. We further show that nerve growth factor (NGF), a known factor of chondrocyte growth, is a key target of DNM3OS-mediated control of chondrocyte proliferation. This work demonstrates that DNM3OS is essential for preventing premature differentiation of chondrocytes required for bone growth through endochondral ossification.
Sigma-Aldrich
Ficoll® 400, BioXtra, for molecular biology, lyophilized powder
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
DAPI ready made solution, For Nuclear counterstain in immunoflourecence microscopy, High Content Screening (HCS), Chromosome staining and flow cytometry (FACS)., 1 mg/mL
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Naphthol AS-MX phosphate, powder, ≥99% (HPLC)