Merck
  • Lightsheet fluorescence lifetime imaging microscopy with wide-field time-correlated single photon counting.
Lightsheet fluorescence lifetime imaging microscopy with wide-field time-correlated single photon counting.
Journal of biophotonics (2019-10-30)
Liisa M Hirvonen, Jakub Nedbal, Norah Almutairi, Thomas A Phillips, Wolfgang Becker, Thomas Conneely, James Milnes, Susan Cox, Stephen Stürzenbaum, Klaus Suhling
ABSTRACT
We report on wide-field time-correlated single photon counting (TCSPC)-based fluorescence lifetime imaging microscopy (FLIM) with lightsheet illumination. A pulsed diode laser is used for excitation, and a crossed delay line anode image intensifier, effectively a single-photon sensitive camera, is used to record the position and arrival time of the photons with picosecond time resolution, combining low illumination intensity of microwatts with wide-field data collection. We pair this detector with the lightsheet illumination technique, and apply it to 3D FLIM imaging of dye gradients in human cancer cell spheroids, and C. elegans.
Sigma-Aldrich
Acridine Orange hydrochloride solution, ≥95.0% (HPLC), 10 mg/mL in H2O
Sigma-Aldrich
Lumidot CdSe/ZnS, 530, core-shell type quantum dots, 5 mg/mL in toluene
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Supelco
Micro particles based on melamine resin, FITC-marked, analytical standard, size: 10 μm