Merck
  • A platform for assessing outer segment fate in primary human fetal RPE cultures.
A platform for assessing outer segment fate in primary human fetal RPE cultures.
Experimental eye research (2018-10-20)
Qitao Zhang, Feriel Presswalla, Kecia Feathers, Xu Cao, Bret A Hughes, David N Zacks, Debra A Thompson, Jason M L Miller
ABSTRACT
The daily shedding and renewal of photoreceptor outer segments (OS) is critical for maintaining vision. This process relies on the efficient uptake, degradation, and sorting of shed OS material by the retinal pigment epithelium (RPE). Poor OS degradation has been linked to retinal degenerations such as Stargardt disease and may contribute to macular degeneration. While primary human fetal RPE cultures have emerged as a valuable model of in vivo human RPE function, surprisingly few studies have utilized the model for tracking the degradation and fate of OS components in the RPE. Here, we establish an improved platform for studying this topic by modifying existing protocols and creating new methods. Our human fetal culture model facilitates studies of RPE secretion in response to OS ingestion, preserves RPE differentiation and polarization during live-cell imaging of OS phagocytosis, and minimizes costs. We optimize Mer tyrosine kinase-dependent OS phagocytosis assays specifically in human fetal cultures and provide a simple and accurate method for measuring total OS consumption by the RPE. Finally, we utilize chemical transfection, dextran labeling, and immunocytochemistry to evaluate key players in OS degradation, including lysosomes and autophagy proteins. To facilitate quantification of autophagy vesicles, we develop customized image analysis macros in the Fiji/ImageJ software environment. These protocols will facilitate a broad range of studies in human fetal RPE cultures aimed at determining the ultimate fate of OS components after ingestion, a critical step in understanding the pathogenesis of numerous retinal diseases.
Sigma-Aldrich
Anti-ZO-1 Antibody, clone R40.76, clone R40.76, from rat