Merck
  • Adenosine A1 and A2A receptors differently control synaptic plasticity in the mouse dorsal and ventral hippocampus.
Adenosine A1 and A2A receptors differently control synaptic plasticity in the mouse dorsal and ventral hippocampus.
Journal of neurochemistry (2019-07-06)
Sara L Reis, Henrique B Silva, Margarida Almeida, Rodrigo A Cunha, Ana P Simões, Paula M Canas
ABSTRACT
The hippocampus is a brain region involved in processing both memory and emotions, through a preferential involvement of the dorsal hippocampus (DH) and ventral hippocampus (VH), respectively. Adenosine A1 and A2A receptors (A1 R and A2A R) control both mood and memory, but it is not known if there is a different adenosine modulation of synaptic plasticity along the hippocampal axis. Using adult, C57BL/6 male mice, we show that both A1 R and A2A R were more abundant in DH compared with VH. However, recordings of field excitatory postsynaptic potentials at Schaffer collaterals-CA1 pyramidal synapses revealed that A1 R were equi-effective to inhibit basal excitatory synaptic transmission in DH and VH, but endogenous A1 R activation was more effective to depress the probability of release in VH. In contrast, the selective A2A R antagonist (SCH58261, 50 nM) controlled both long-term potentiation (induced by a high frequency stimulation protocol) and long-term depression (induced by a low frequency stimulation protocol) selectively in DH rather than VH, whereas the selective A1 R antagonist (DPCPX, 100 nM) revealed a similar tonic inhibition of long-term depression in DH and VH. These findings show a different control of synaptic plasticity by the adenosine modulation system in the dorsal and ventral poles of the hippocampus, which may underlie a different efficiency of the adenosine system to control mood and memory.
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Roche
Adenosine Deaminase (ADA), from calf intestine
Sigma-Aldrich
Methyl sulfoxide, ≥99%, FG
Sigma-Aldrich
2-Chloroadenosine