Merck
  • TREM2 Regulates Microglial Cholesterol Metabolism upon Chronic Phagocytic Challenge.
TREM2 Regulates Microglial Cholesterol Metabolism upon Chronic Phagocytic Challenge.
Neuron (2020-01-07)
Alicia A Nugent, Karin Lin, Bettina van Lengerich, Steve Lianoglou, Laralynne Przybyla, Sonnet S Davis, Ceyda Llapashtica, Junhua Wang, Do Jin Kim, Dan Xia, Anthony Lucas, Sulochanadevi Baskaran, Patrick C G Haddick, Melina Lenser, Timothy K Earr, Ju Shi, Jason C Dugas, Benjamin J Andreone, Todd Logan, Hilda O Solanoy, Hang Chen, Ankita Srivastava, Suresh B Poda, Pascal E Sanchez, Ryan J Watts, Thomas Sandmann, Giuseppe Astarita, Joseph W Lewcock, Kathryn M Monroe, Gilbert Di Paolo
ABSTRACT
Loss-of-function (LOF) variants of TREM2, an immune receptor expressed in microglia, increase Alzheimer's disease risk. TREM2 senses lipids and mediates myelin phagocytosis, but its role in microglial lipid metabolism is unknown. Combining chronic demyelination paradigms and cell sorting with RNA sequencing and lipidomics, we find that wild-type microglia acquire a disease-associated transcriptional state, while TREM2-deficient microglia remain largely homeostatic, leading to neuronal damage. TREM2-deficient microglia phagocytose myelin debris but fail to clear myelin cholesterol, resulting in cholesteryl ester (CE) accumulation. CE increase is also observed in APOE-deficient glial cells, reflecting impaired brain cholesterol transport. This finding replicates in myelin-treated TREM2-deficient murine macrophages and human iPSC-derived microglia, where it is rescued by an ACAT1 inhibitor and LXR agonist. Our studies identify TREM2 as a key transcriptional regulator of cholesterol transport and metabolism under conditions of chronic myelin phagocytic activity, as TREM2 LOF causes pathogenic lipid accumulation in microglia.
Sigma-Aldrich
GW3965 hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Avanti
18:0-20:4 PI, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse