Merck
  • Heme oxygenase-1 attenuates seawater drowning-induced acute lung injury through a reduction in inflammation and oxidative stress.
Heme oxygenase-1 attenuates seawater drowning-induced acute lung injury through a reduction in inflammation and oxidative stress.
International immunopharmacology (2019-06-30)
Xue-Qian Sun, Chen Wu, Yu-Bao Qiu, Ya-Xian Wu, Jun-Liang Chen, Jian-Feng Huang, Dan Chen, Qing-Feng Pang
ABSTRACT
Heme oxygenase-1 (HO-1) plays a critical protective role in various insults-induced acute lung injury (ALI) through its strong anti-inflammatory, anti-oxidant, and anti-apoptotic properties, but its protective role and mechanism on seawater aspiration-induced acute lung injury remains unclear. This study aimed to explore the therapeutic potential and mechanism of HO-1 to attenuate seawater aspiration-induced ALI in vivo and in vitro. The viability and invasion of A549 cell were analyzed through cell counting kit-8 and lactate dehydrogenase release assay; the transcriptional level of inflammatory cytokines (TNF-α, IL-6, IL-8 and MCP-1) and cell proliferation-related cytokines (FoxM1, Ccnb1 and Cdc25C) in seawater-treated A549 cell were tested by qPCR; apoptotic cells were analyzed by flow cytometryd; HO-1mRNA and protein were determined by qPCR and western blotting; the fluorescent indicators (DCFH-DA, dihydroethidium, MitoSox Red and Fluo-4) were used to monitor generation of ROS and mitochondrial function. The lung wet/dry weight radio and lactate dehydrogenase activity, Sirius red staining, TUNEL assay and immunohistochemical staining with anti-pan Cytokeratin antibody were analyzed in seawater-drowning mice. The role of HO-1 on seawater-drowning pulmonary injury was explored via HO-1 activity inhibitors (Zinc protoporphyrin) in vitro and in vivo. Seawater exposure decreased the cellular viability, increased the production of pro-inflammatory cytokines (IL-6, IL-8 and TNF-α), induced cellular apoptosis and inhibited the expression of cell proliferation-related cytokines (FoxM1, Ccnb1 and Cdc25C). Moreover, seawater exposure led to mitochondrial dysfunction in A549 cells. Supplement of HO-1 sepcific inducer (heme) or its catalytic product (biliverdin) significantly attenuated seawater-induced A549 damage and promoted cell proliferation. However, Zinc protoporphyrin abolished the beneficial effects of HO-1 on seawater drowning-induced pulmonary tissue injury. HO-1 attenuates seawater drowning-induced lung injury by its anti-inflammatory, anti-oxidative, and anti-apoptosis function.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate, reduced tetra(cyclohexylammonium) salt, ≥93%
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Biliverdin hydrochloride, ≥97.0% (TLC)