Merck
  • Extracellular Matrix Components HAPLN1, Lumican, and Collagen I Cause Hyaluronic Acid-Dependent Folding of the Developing Human Neocortex.
Extracellular Matrix Components HAPLN1, Lumican, and Collagen I Cause Hyaluronic Acid-Dependent Folding of the Developing Human Neocortex.
Neuron (2018-08-07)
Katherine R Long, Ben Newland, Marta Florio, Nereo Kalebic, Barbara Langen, Anna Kolterer, Pauline Wimberger, Wieland B Huttner
ABSTRACT
Neocortical expansion, thought to underlie the cognitive traits unique to humans, is accompanied by cortical folding. This folding starts around gestational week (GW) 20, but what causes it remains largely unknown. Extracellular matrix (ECM) has been previously implicated in neocortical expansion and here we investigate the potential role of ECM in the formation of neocortical folds. We focus on three specific ECM components localized in the human fetal cortical plate (CP): hyaluronan and proteoglycan link protein 1 (HAPLN1), lumican and collagen I (collectively, HLC). Addition of HLC to cultures of human fetal neocortex (11-22 GW) caused local changes in tissue stiffness, induced CP folding, increased CP hyaluronic acid (HA), and required the HA-receptor CD168 and downstream ERK signaling. Importantly, loss of HA reduced HLC-induced and 22 GW physiological nascent folds. This was altered in samples with neurodevelopmental disorders, indicating it may be a useful system to study such disorders.
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, BSA free, lyophilized powder, specific activity 50-250 units/mg protein
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid