Merck
  • The chemokine SDF-1, stromal cell-derived factor 1, attracts early stage B cell precursors via the chemokine receptor CXCR4.
The chemokine SDF-1, stromal cell-derived factor 1, attracts early stage B cell precursors via the chemokine receptor CXCR4.
European journal of immunology (1997-07-01)
M D'Apuzzo, A Rolink, M Loetscher, J A Hoxie, I Clark-Lewis, F Melchers, M Baggiolini, B Moser
ABSTRACT
In the bone marrow, progenitor (pro-) and precursor (pre-) B cells depend on close contact with stromal cells for growth and maturation. Stromal cell-derived factor 1 (SDF-1), also known as pre-B cell growth-stimulating factor, is produced by bone marrow stromal cells and was reported to act together with interleukin-7 as co-mitogen for pre-B cells. SDF-1 was recently shown to be a chemokine which is chemotactic for different types of leukocytes and acts via the chemokine receptor CXCR4. Using sorted B220+ bone marrow cells and several B cell lines characteristic for different stages of B lymphopoiesis, we now show that SDF-1 is a potent attractant for pro- and pre-B cells, but is inactive on B cells at later stages of development. In early B cell precursors, SDF-1 induced intracellular Ca2+ mobilization and in vitro migration with a potency and efficacy similar to that observed for chemokines acting on blood leukocytes. These responses were mediated via CXCR4 as they could be inhibited by an antireceptor antibody. SDF-1 is the first chemokine shown to act on early-stage B cell precursors. Mice lacking SDF-1 die perinatally and show a severe deficiency in B lymphopoiesis. We propose that SDF-1 released from the stromal cells exerts its critical hematopoietic function by selectively attracting and confining early B cell precursors within the bone marrow microenvironment that provides the necessary factors for growth and differentiation.
Sigma-Aldrich
SDF-1 BETA human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
SDF-1 alpha from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
SDF-1 beta from rat, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
SDF-1 ALPHA human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)