Merck
Molecular detection of Rickettsia typhi in cats and fleas.
PloS one (2013-08-14)
Maria Mercedes Nogueras, Immaculada Pons, Ana Ortuño, Jaime Miret, Julia Pla, Joaquim Castellà, Ferran Segura
ABSTRACT
Rickettsiatyphi is the etiological agent of murine typhus (MT), a disease transmitted by two cycles: rat-flea-rat, and peridomestic cycle. Murine typhus is often misdiagnosed and underreported. A correct diagnosis is important because MT can cause severe illness and death. Our previous seroprevalence results pointed to presence of human R. typhi infection in our region; however, no clinical case has been reported. Although cats have been related to MT, no naturally infected cat has been described. The aim of the study is to confirm the existence of R. typhi in our location analyzing its presence in cats and fleas. 221 cats and 80 fleas were collected from Veterinary clinics, shelters, and the street (2001-2009). Variables surveyed were: date of collection, age, sex, municipality, living place, outdoor activities, demographic area, healthy status, contact with animals, and ectoparasite infestation. IgG against R. typhi were evaluated by indirect immunofluorescence assay. Molecular detection in cats and fleas was performed by real-time PCR. Cultures were performed in those cats with positive molecular detection. Statistical analysis was carried out using SPSS. A p < 0.05 was considered significant. Thirty-five (15.8%) cats were seropositive. There were no significant associations among seropositivity and any variables. R. typhi was detected in 5 blood and 2 cultures. High titres and molecular detection were observed in stray cats and pets, as well as in spring and winter. All fleas were Ctenocephalides felis. R. typhi was detected in 44 fleas (55%), from shelters and pets. Co-infection with R. felis was observed. Although no clinical case has been described in this area, the presence of R. typhi in cats and fleas is demonstrated. Moreover, a considerable percentage of those animals lived in households. To our knowledge, this is the first time R. typhi is detected in naturally infected cats.
Sigma-Aldrich
Anti-Cat IgG (whole molecule)−FITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution