Merck
  • Laboratory validation of an LC-MS/MS method for the detection of ractopamine, clenbuterol and salbutamol in bovine and swine muscle at sub-μg kg
Laboratory validation of an LC-MS/MS method for the detection of ractopamine, clenbuterol and salbutamol in bovine and swine muscle at sub-μg kg
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-03-10)
Alfredo M Montes Nino, Rodrigo H M M Granja, Karine V G Reche, Fabio M Giannotti, João K G de Souza, Sarita P G Ferrari, André D Dos Santos, Amarylis C B A Wanschel, Alessandro G Salerno
ABSTRACT
Ractopamine (RAC), is a β-adrenergic agonist increasingly used in the swine and cattle industry. This compound redirects nutrients to favour leanness rather than fat deposition, improves growth and carcass traits gaining higher economic benefit to producers. Countries around the world are split over whether to allow the use of RAC in meat production. Clenbuterol (CLB) and salbutamol (SLB) are anillinic and phenolic β-agonists, respectively, with the same capacity of producing economic benefits for the meat sector. However, they are prohibited because of the potentially adverse reactions they can cause in consumers. The three β-agonist compounds have been included in the Brazilian National Regulatory Survey and consequentially there is an eminent need for reliable methods capable of detecting those substances at the same time and reduce analytical costs. Therefore, an LC-MS/MS method for the simultaneous determination of residual RAC, CLB and SAL in swine and cattle muscle was developed and validated with quantification levels respecting the action levels established for Brazil which are 0.1, 0.2 and 5 µg kg
Product Description
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.10 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.10 μm
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 20 m × 0.18 mm, df 0.18 μm