
Wang resin

Synonym(s):
p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene
Products
Product Category
Brand
Physical Form
Quality Segment
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-3 of 3