Skip to Content
Merck
Trioctylamine 98%

Trioctylamine

Trioctylamine 98%
Synonym(s):
TOA
Linear Formula:
[CH3(CH2)7]3N
CAS Number:
Molecular Weight:
353.67
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing