Skip to Content
Merck
Trimethyl orthoformate anhydrous, 99.8%

Trimethyl orthoformate

Trimethyl orthoformate anhydrous, 99.8%
Synonym(s):
Trimethoxymethane
Linear Formula:
CH(OCH3)3
CAS Number:
Molecular Weight:
106.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing