
Trimethoxyphenylsilane

Synonym(s):
(Trimethoxysilyl)benzene, Phenyltrimethoxysilane
Linear Formula:
C6H5Si(OCH3)3
CAS Number:
Molecular Weight:
198.29
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing