
Theaflavin 3,3′-digallate

Synonym(s):
3,4,5-Trihydroxy-benzoic acid (3,4,6-trihydroxy-5-oxo-5H-benzocycloheptene-1,8-diyl)bis[(2R,3R)-3,4-dihydro-5,7-dihydroxy-2H-1-benzopyran-2,3-diyl] ester, [3-hydroxy-5-oxo-4-(3,4,5-trihydroxybenzoyl)oxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-3,4-dihydro-2H-chromen-2-yl]benzo[7]annulen-6-yl] 3,4,5-trihydroxybenzoate
Empirical Formula (Hill Notation):
C43H32O20
CAS Number:
Molecular Weight:
868.70
Products
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing