
Sulfochlorophenol S sodium calcium salt

Synonym(s):
2,7-Bis(2-hydroxy-3-sulfo-5-chlorophenylazo)-1,8-di-hydroxynaphthalene-3,6-disulfonic acid
Empirical Formula (Hill Notation):
C22H14Cl2N4O16S4
CAS Number:
Molecular Weight:
789.53
Products
Product Category
Brand
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing